Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Folic acid, Clone: FA3, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10072977
Description
Folic acid Monoclonal specifically detects Folic acid in Non-species specific samples. It is validated for Enzyme Immunoassay.Specifications
Folic acid | |
Monoclonal | |
Unconjugated | |
5 Methyl THFA, 5 Methyltetrahydrofolate, Folate, Tetrahydrofolate, Tetrahydrofolic acid, THFA | |
Derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with folic acid conjugated with BSA. | |
0.2 mg | |
Primary | |
Non-species specific | |
Purified |
ELISA | |
FA3 | |
Enzyme Immunoassay | |
Mouse | |
Protein G purified | |
RUO | |
This recognizes free folic acid. | |
Store at 4C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction