Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP3 Antibody (3G3), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056582V
Description
FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
| FoxP3 | |
| Monoclonal | |
| DyLight 405 | |
| Q9BZS1 | |
| FOXP3 | |
| Full-length His-tagged murine FOXP3 was used as the immunogen. The precise epitope is not known, but it has been mapped to the N-terminal portion of the protein (NP_001107849). | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry | |
| 3G3 | |
| Western Blot, Flow Cytometry | |
| AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
| Mouse | |
| 47 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Transcription Factors and Regulators | |
| 50943 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction