Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2AX, p Ser139 Antibody (9F3), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP119255
Description
Histone H2AX (p Ser139) Monoclonal antibody specifically detects Histone H2AX (p Ser139) in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
Histone H2AX (p Ser139) | |
Monoclonal | |
Unconjugated | |
P16104 | |
H2AFX | |
This Histone H2AX [p Ser139] Antibody (9F3) was developed against a synthetic phospho-peptide derived from the sequence of human Histone H2AX phosphorylated at Ser139. | |
Protein G purified | |
RUO | |
Primary | |
Reacts with Monkey, Hamster. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
9F3 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunocytochemistry | |
H2A.X, H2A/X, H2AFX | |
Mouse | |
15 kDa | |
0.05 mg | |
Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific | |
3014 | |
Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Guinea Pig, Hamster, Primate, Rabbit, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction