Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFAP Mouse anti-Human, Mouse, Rat, Monkey, DyLight 350, Clone: GF5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010062UV
Description
GFAP Monoclonal specifically detects GFAP in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GFAP | |
| Monoclonal | |
| DyLight 350 | |
| FLJ45472, GFAP astrocytes, glial fibrillary acidic protein | |
| Mouse | |
| 50 kDa | |
| 0.2 mL | |
| Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 2670 | |
| Human, Mouse, Rat, Monkey | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| GF5 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| GFAP | |
| The GFAP Antibody (GF5) hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with native human GFAP. | |
| Protein G purified | |
| RUO | |
| Primary | |
| There is no cross-reactivity with other neurospecific proteins. | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction