Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-2Dd Antibody (34-5-8S), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128186
Description
H-2Dd Monoclonal specifically detects H-2Dd in Mouse samples. It is validated for Flow Cytometry.Specifications
H-2Dd | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry 1 ug/10^6 cells | |
Mouse | |
Protein A or G purified | |
RUO | |
A conformational epitope on the alpha1 and alpha2 chains of mouse H-2Dd class I alloantigen | |
Store at 4C. Do not freeze. | |
IgG2a κ |
Flow Cytometry, Immunoprecipitation, Blocking Assay, Immunocytochemistry | |
34-5-8S | |
FITC | |
H2Dd, MHC class I | |
BDF1 mouse splenocytes | |
0.25 mg | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction