Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HSP10/EPF, DyLight 488, Clone: M1.2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12013527G
Description
HSP10/EPF Monoclonal specifically detects HSP10/EPF in Bacteria samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| HSP10/EPF | |
| Monoclonal | |
| DyLight 488 | |
| 10 kDa chaperonin, 10 kDa heat shock protein, mitochondrial, Chaperonin 10, CPN10HSP10, Early-pregnancy factor, EPF, GROES, heat shock 10kD protein 1 (chaperonin 10), heat shock 10kDa protein 1 (chaperonin 10), Hsp10 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This affinity purified detects a 15 kDa protein, corresponding to the apparent molecular mass of cpn10 on SDS-PAGE immunoblots, in C. trachomatis infected cells. This will not react with the human or mouse cpn10 homolog nor with E. coli GroES. | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| M1.2 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| HSPE1 | |
| A 16 residue synthetic MAP peptide KKDDKGQQLPFEVQVG based on C. trachomatis cpn10 (residues 54-69). | |
| 0.1 mL | |
| Core ESC Like Genes, Membrane Trafficking and Chaperones, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| 3336 | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction