Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IGFBP-4, Clone: IBP163, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP232773
Description
IGFBP-4 Monoclonal specifically detects IGFBP-4 in Human, Mouse samples. It is validated for ELISA, Sandwich ELISA.Specifications
IGFBP-4 | |
Monoclonal | |
Unconjugated | |
BP-4, HT29-IGFBP, IBP4insulin-like growth factor-binding protein 4, IGF-binding protein 4, IGFBP-4IBP-4, insulin-like growth factor binding protein 4 | |
Mouse | |
Protein A purified | |
RUO | |
3487 | |
Store at 4C. Do not freeze. | |
IgG1 |
ELISA | |
IBP163 | |
ELISA 1:100-1:2000, Sandwich ELISA 1:100-1:2000 | |
IGFBP4 | |
IGFBP-4 | |
0.2 mg | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction