Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse IL-17 RC, Fluorescein, Polyclonal, R&D Systems™

Catalog No. FAB2270F
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2270F 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2270F Supplier R&D Systems Supplier No. FAB2270F
This item is not returnable. View return policy

Host Species: Goat
Species Reactivity:Mouse
Immunogen:Mouse myeloma cell line NS0-derived recombinant mouse IL&No.x2011;17&No.160;RC, Leu21-Trp465, Accession No. AAH04759

Applications:

Flow Cytometry

Specifications

Antigen IL-17 RC
Applications Flow Cytometry
Classification Polyclonal
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. AAH04759
Gene Alias FLJ95963, FLJ96005, IL-17 receptor C, IL-17 RL, IL17F receptor, IL17RC, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14
Gene Symbols IL17RC
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RC, Leu21-Trp465, Accession # AAH04759
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84818
Test Specificity Detects mouse IL-17 RC in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human IL-17 RC is observed and less than 5% cross-reactivity with recombinant mouse (rm) IL-17 RD and rmIL-17B R is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.