Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17F Antibody (4H450), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227319B
Description
IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for Western Blot.Specifications
| IL-17F | |
| Monoclonal | |
| Biotin | |
| Q96PD4 | |
| IL17F | |
| Full-length recombinant MRkIPkVGHT FFQkPESCPP VPGGSMkLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGkEDISMN SVPIQQETLV VRRkHQGCSV SFQLEkVLVT VGCTCVTPVI HHVQ | |
| 0.1 mL | |
| Cytokine Research | |
| 112744 | |
| Human | |
| Purified |
| Western Blot | |
| 4H450 | |
| Western Blot | |
| Cytokine ML-1, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is reactive to Human. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction