Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 3/CD61 Antibody (BV4), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 7 publications
Supplier: Novus Biologicals NB6001342
Description
Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.Specifications
Integrin beta 3/CD61 | |
Monoclonal | |
0.1 mg/mL | |
Western Blot 1:50, Immunohistochemistry 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:50, Functional, Immunoassay | |
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
The monoclonal antibody BV4 recognizes human beta3 integrin subunit present in Platelet glycoprotein GPIIb-IIIa (integrin alphaIIb/beta3, CD41/CD61) and in the vitronectin receptor (integrin alphaV/beta3, CD51/CD61). | |
Store at 4°C. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Immunoassay | |
BV4 | |
Unconjugated | |
P05106 | |
ITGB3 | |
Mouse Integrin alpha V beta 3 | |
0.05 mg | |
Phospho Specific | |
3690 | |
Human, Mouse, Rat, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction