Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Lactoferrin, Clone: 2B8, Novus Biologicals™

Mouse Monoclonal Antibody
$122.00
Specifications
Antigen | Lactoferrin |
---|---|
Clone | 2B8 |
Dilution | Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB6001529SS
![]() |
Novus Biologicals
NB6001529SS |
0.02 mg |
Each of 1 for $122.00
|
|
|||||
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Western Blot, Sandwich ELISA.Specifications
Lactoferrin | |
Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000 | |
Monoclonal | |
Mouse | |
Amino Acids Drugs and other small molecules | |
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
LTF | |
IgG1 κ | |
Affinity Purified | |
Does not cross-react with human transferrin or other serum proteins. |
2B8 | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Human | |
4057 | |
Full length native protein (purified) (Human). | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title