Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDLR Antibody (C7), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 19 publications
$206.50 - $501.50
Specifications
Antigen | LDLR |
---|---|
Clone | C7 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Flow Cytometry reported in scientific literature (PMID 10906332), ELISA, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 11839845), In vitro assay reported in scientific literature (PMID 9642270), Radioimmunoassay, Proximity Ligation Assay reported in scientific literature (PMID 3263645), Electron Microscopy reported in scientific literature (PMID 11839845) |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, Radioimmune Assays (RIA), In situ PLA, Immunoassay |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP178159SS
![]() |
Novus Biologicals
NBP178159SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP178159
![]() |
Novus Biologicals
NBP178159 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
LDLR Monoclonal specifically detects LDLR in Human, Rat, Porcine, Bovine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Radioimmunoassay, Proximity Ligation Assay, Electron Microscopy.Specifications
LDLR | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, Radioimmune Assays (RIA), In situ PLA, Immunoassay | |
Unconjugated | |
Mouse | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
P01130, P01131 | |
3949 | |
Partially purified bovine LDL receptor [Swiss-Prot# P01131]. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
C7 | |
Western Blot 1:1000, Flow Cytometry reported in scientific literature (PMID 10906332), ELISA, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 11839845), In vitro assay reported in scientific literature (PMID 9642270), Radioimmunoassay, Proximity Ligation Assay reported in scientific literature (PMID 3263645), Electron Microscopy reported in scientific literature (PMID 11839845) | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Pig, Bovine | |
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
LDLR | |
IgG2b | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title