Learn More
LPAM-1 Rat anti-Mouse, PE-CF594, Clone: DATK32, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 562668
Description
The DATK32 monoclonal antibody specifically binds to an epitope specific for the α4β7 integrin heterodimer. α4β7 (LPAM-1) is expressed on most mature lymphocytes and on small subsets of thymic and bone marrow cells. It interacts with several ligands, including VCAM-1 (CD106), fibronectin, and MAdCAM-1. DATK32antibody induces α4β7-dependent lymphocyte aggregation, but it inhibits other α4β7- mediated lymphocyte adhesion events, including binding to fibronectin, MAdCAM-1, and VCAM-1 (CD106).
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

Specifications
LPAM-1 | |
Monoclonal | |
0.2mg/mL | |
Itga4Itgb7; α4β7 Integrin; alpha 4 beta 7 integrin; CD49dβ7 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
DATK32 | |
PE-CF594 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
AKR/Cum mouse T lymphoma line TK1 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.