Learn More
Description
Host Species: Rat
Clone: D7
Isotype: IgG2a, κ
Cross Reactivity Species: Ms
Immunogen: IL-2-dependent mouse T-cell line CTL-L
Flow Cytometry
Specifications
Specifications
| Antigen | Mouse Ly-6A/E |
| Classification | Monoclonal |
| Clone | D7 |
| Concentration | 0.2mg/mL |
| Conjugate | PE-CF594 |
| Host Species | Rat |
| Immunogen | IL-2-dependent mouse T-cell line CTL-L |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.