Learn More
Description
Host Species: Rat
Clone: 1A8
Isotype: IgG2a, κ
Cross Reactivity Species: Ms
Immunogen: Ly-6G-transfected EL4J cell line
Flow Cytometry
Specifications
Specifications
| Antigen | Mouse LY-6G |
| Classification | Monoclonal |
| Clone | 1A8 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 421 |
| Host Species | Rat |
| Immunogen | Ly-6G-transfected EL4J cell line |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.