Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse LYVE-1 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF2125 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2125 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2125 Supplier R&D Systems Supplier No. AF2125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 49 publications

LYVE-1 Polyclonal specifically detects LYVE-1 in Human∣Mouse∣Pig∣Rat∣Transgenic Mouse samples. It is validated for Cell Selection∣Dual RNAScope ISH-IHC Compatible∣Flow Cytometry∣IF/IHC∣Immunocytochemistry∣Immunocytochemistry/Immunofluorescence∣Immunofluorescence∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Immunoprecipitation∣Western Blot.

Specifications

Antigen LYVE-1
Applications Cell Selection, Dual RNAScope ISH-IHC, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q8BHC0
Gene Alias cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1XLKD1, XLKD1
Gene Symbols LYVE1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse LYVE-1 Ala24-Thr234 Accession # Q8BHC0
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10894
Test Specificity Detects mouse LYVE-1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Pig, Rat, Transgenic Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.