Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-MBD3/MBD2, Clone: 106B691, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$182.00 - $464.00
Specifications
Antigen | MBD3 |
---|---|
Clone | 106B691 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 ug/ml, Chromatin Immunoprecipitation, Flow Cytometry 1 ug million cells, Immunohistochemistry-Paraffin, Immunocytochemistry |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056534T
![]() |
Novus Biologicals
NB10056534SS |
0.025 mg |
Each for $182.00
|
|
|||||
NB10056534
![]() |
Novus Biologicals
NB10056534 |
0.1 mg |
Each for $464.00
|
|
|||||
Description
MBD3 Monoclonal specifically detects MBD3 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.Specifications
MBD3 | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Mouse | |
Chromatin Research | |
O95983 | |
53615 | |
Between amino acids 200-250 of human MBD3 protein were used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
106B691 | |
Western Blot 2 ug/ml, Chromatin Immunoprecipitation, Flow Cytometry 1 ug million cells, Immunohistochemistry-Paraffin, Immunocytochemistry | |
Monoclonal | |
Purified | |
RUO | |
Human | |
methyl-CpG binding domain protein 3, methyl-CpG-binding domain protein 3, Methyl-CpG-binding protein MBD3 | |
MBD3 | |
IgG1 κ | |
Protein G purified | |
The theoretical molecular weight of human MBD2 (411 amino acids) is 45 kDa and human MBD3 (291 amino acids) is 33 kDa. Based on sequence identity, this will detect human, mouse, and rat MBD3 (100%), and human and mouse MBD2 (93%). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title