Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCTS1 Antibody (OTI2F9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP203626
Description
MCTS1 Monoclonal specifically detects MCTS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MCTS1 | |
| Monoclonal | |
| 0.81 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
| MCTS1 | |
| Human recombinant protein fragment corresponding to amino acids 1-181 of human MCTS1 (NP_054779) produced in E.coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI2F9 | |
| Unconjugated | |
| malignant T cell amplified sequence 1, Malignant T-Cell-Amplified Sequence 1, MCT-1, Multiple Copies T-Cell Malignancies | |
| Mouse | |
| 20.4 kDa | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 28985 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction