Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIST1 Antibody (6E8/A12/C11P1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222478
Description
MIST1 Monoclonal specifically detects MIST1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| MIST1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:500, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| basic helix-loop-helix domain containing, class B, 8, basic helix-loop-helix family, member a15, bHLHa15, bHLHb8, class A basic helix-loop-helix protein 15, Class B basic helix-loop-helix protein 8, class II bHLH protein MIST1, MIST1MIST-1, Muscle, intestine and stomach expression 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 168620 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 6E8/A12/C11P1 | |
| Unconjugated | |
| Q7RTS1 | |
| BHLHA15 | |
| Mouse Mist1, C-terminus residues 175-197. | |
| 100 μg | |
| Primary | |
| This antibody reacts with Human and Mouse. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction