Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Cadherin Mouse anti-Human, Mouse, Rat, Clone: GC-4, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12011340
Description
N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
N-Cadherin | |
Monoclonal | |
Unconjugated | |
cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin | |
Mouse | |
135 kDa | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells | |
1000 | |
Human, Mouse, Rat, Chicken, Rabbit | |
Ascites |
Western Blot, Immunocytochemistry, Immunofluorescence | |
GC-4 | |
Western Blot 1:100, Immunocytochemistry/Immunofluorescence 1:100 | |
CDH2 | |
This N-Cadherin Antibody (GC-4) - Unpurified was developed against recombinant full length protein (Chicken heart). | |
Unpurified | |
RUO | |
Primary | |
This N-Cadherin Antibody (GC-4) - Unpurified reacts with the N-terminal portion of the extracellular domain of A-CAM. It inhibits adherens junction formation and disrupts existing junctions of cultured lens cells. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction