Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Nesfatin-1/Nucleobindin-2, Clone: Nesfaty-1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197633
Description
Nesfatin-1/Nucleobindin-2 Monoclonal specifically detects Nesfatin-1/Nucleobindin-2 in Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
Nesfatin-1/Nucleobindin-2 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:100 | |
NUCB2 | |
Recombinant rat nesfatin-1 (aa 25-106). | |
0.05 mg | |
Primary | |
Recognizes mouse and rat nesfatin-1 and precursor nucleobindin-2. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunoprecipitation | |
Nesfaty-1 | |
Unconjugated | |
DNA-binding protein NEFA, NEFAGastric cancer antigen Zg4, nucleobindin 2, nucleobindin-2, nucleobinding 2 | |
Mouse | |
Protein G purified | |
RUO | |
4925 | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction