Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-H Antibody (RNF402), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197726
Description
NF-H Monoclonal specifically detects NF-H in Human, Mouse, Rat, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
NF-H | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:25-1:250, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:100, Immunohistochemistry-Frozen 1:10-1:100, Immunoblotting 1:25-1:250 | |
NEFH | |
RNF402 is a mouse monoclonal IgM antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a neurofilament preparation of calf brain tissue. | |
0.1 mg | |
Autophagy, Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience | |
4744 | |
Human, Mouse, Rat, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep, Xenopus species | |
IgM |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
RNF402 | |
Unconjugated | |
200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
RNF402 reacts with both the phosphorylated and non-phosphorylated isoform of the 200 kD neurofilament protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction