Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-M Antibody (NF-09), Alexa Fluor™ 405

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500446AF405
Description
NF-M Monoclonal specifically detects NF-M in Mouse, Porcine, Mammal samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NF-M | |
Monoclonal | |
Alexa Fluor 405 | |
NEF3neurofilament medium polypeptide, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The antibody NF-09 reacts with both phosphorylated and non-phosphorylated form of medium neurofilament protein (160 kDa) of various species. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
NF-09 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
NEFM | |
Pellet of pig brain cold stable proteins after depolymerization of microtubules. | |
0.1 mL | |
Autophagy, Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
4741 | |
Mouse, Pig, Mammalia | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction