Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse OX40 Ligand/TNFSF4, Fluorescein, Polyclonal, R&D Systems™

Catalog No. FAB1236F
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1236F 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1236F Supplier R&D Systems Supplier No. FAB1236F
This item is not returnable. View return policy

Host Species: Goat
Species Reactivity:Mouse
Immunogen:Mouse myeloma cell line NS0-derived recombinant mouse OX40 Ligand/TNFSF4, Gln49-Leu198, Accession No. P43488

Applications:

Flow Cytometry

Specifications

Antigen OX40 Ligand/TNFSF4
Applications Flow Cytometry
Classification Polyclonal
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P43488
Gene Alias CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1
Gene Symbols TNFSF4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse OX40 Ligand/TNFSF4, Gln49-Leu198, Accession # P43488
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7292
Test Specificity Detects mouse OX40 Ligand/TNFSF4 in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human (rh) OX40 Ligand, rhLIGHT, recombinant mouse (rm) Fas Ligand and less than 1% cross-reactivity with rmTNF-alpha , rhGITR Ligand, rhTRANCE, and rhVEGI is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.