Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRC1 Antibody (16F2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10074508
Description
PRC1 Monoclonal specifically detects PRC1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
PRC1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1 - 3 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunoprecipitation 1:10 - 1:500 | |
anaphase spindle elongation 1 homolog, ASE1, MGC1671, MGC3669, protein regulating cytokinesis 1, protein regulator of cytokinesis 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
PRC1 (16F2) | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
16F2 | |
Unconjugated | |
O43663 | |
PRC1 | |
Recombinant PRC1 protein. | |
100 μg | |
Cell Cycle and Replication, Mitotic Regulators | |
9055 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction