Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prox1 Antibody (5G10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP130045
Description
Prox1 Monoclonal specifically detects Prox1 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Whole-Mount, CyTOF-ready.Specifications
Prox1 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry Whole-Mount reported in scientific literature (PMID 24654984), CyTOF-ready | |
Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Specific for Prox1. It is expected to have wide species cross reactivity. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
5G10 | |
Unconjugated | |
Q92786, Q92786, P48437 | |
PROX1 | |
The highly conserved Homeo and Prospero domain of human Prox1. [UniProt# Q92786] | |
0.1 mL | |
Vision | |
5629 | |
Human, Mouse, Rat, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction