Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R9AP Antibody (D7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222470
Description
R9AP Monoclonal specifically detects R9AP in Mouse, Bovine samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.Specifications
R9AP | |
Monoclonal | |
1.93 mg/mL | |
Western Blot 1 ug/ml, Immunoprecipitation 1:10 - 1:500, Immunocytochemistry 10 ug/ml | |
RGS9BP | |
Full length, bacterially expressed RGS9-1. | |
Protein G purified | |
RUO | |
388531 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunoprecipitation, Immunocytochemistry | |
D7 | |
Unconjugated | |
PERRS, R9AP, regulator of G protein signaling 9 binding protein, RGS9 | |
Mouse | |
57 kDa | |
100 μg | |
Primary | |
Mouse, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction