Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Rabies Virus, Clone: RV1C5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1102523
Description
Rabies Virus Monoclonal specifically detects Rabies Virus in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence, Enzyme Immunoassay.Specifications
Rabies Virus | |
Monoclonal | |
Unconjugated | |
Glycoprotein | |
Purified rabies virus. | |
0.2 mg | |
Virology Bacteria and Parasites | |
This reacts with a glycoprotein of rabies virus. More than 20 different strains from 4 serogroups, including CVS, Lagosbat, Mokola, Duwenhage were positive in neutralisation reaction with this. | |
Store at 4C. Do not freeze. | |
IgG2a |
Immunocytochemistry, Immunofluorescence, ELISA | |
RV1C5 | |
Immunocytochemistry/Immunofluorescence 1:10-1:500, Enzyme Immunoassay | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction