Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A8/A9 Antibody (48M7C7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP225269H
Description
S100A8/A9 Monoclonal specifically detects S100A8/A9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
S100A8/A9 | |
Monoclonal | |
HRP | |
60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8, S100 calcium binding protein A8, S100A8 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This product reacts with Human and Mouse. | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
48M7C7 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
S100A8 | |
Within the range of amino acids 2-105 of human S100A8 were used as the immunogen for the antibody. | |
0.1 mL | |
Cancer | |
6279 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction