Learn More
Description
Host Species: Rat
Clone: E50-2440
Isotype: IgG2a, κ
Cross Reactivity Species: Ms
Immunogen: Mouse Siglec-F and human IgG Fc recombinant fusion protein
Flow Cytometry
Specifications
Specifications
| Antigen | Mouse Siglec-F |
| Classification | Monoclonal |
| Clone | E50-2440 |
| Concentration | 0.2mg/mL |
| Conjugate | PE-CF594 |
| Host Species | Rat |
| Immunogen | Mouse Siglec-F and human IgG Fc recombinant fusion protein |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.