Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Staphylococcus aureus Enterotoxin Type A, Clone: C4, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11059900
Description
Staphylococcus Aureus Enterotoxin Type A Monoclonal specifically detects Staphylococcus Aureus Enterotoxin Type A in Non-species specific samples. It is validated for Western Blot, ELISA, Sandwich ELISA.Specifications
Staphylococcus Aureus Enterotoxin Type A | |
Monoclonal | |
Unconjugated | |
Ent A, EntA, Enterotoxin A, Enterotoxin type A, SEA, Staphylococcus enterotoxin type A | |
KLH-PEG with terminal methoxy group was used as an immunogen. This recognizes the terminal methoxy group of the PEG molecule. Anti-PEG-47-Biotin is prepared by the biotinylation of the via primary amines. | |
0.2 mg | |
Primary | |
Non-species specific | |
Purified |
Western Blot, ELISA | |
C4 | |
Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000 | |
Mouse | |
Protein G purified | |
RUO | |
Staphylococcus aureus Enterotoxin type A (C4) | |
Store at 4C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction