Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIRAP Antibody (20D1055.1), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen TIRAP
Clone 20D1055.1
Dilution Western Blot 1:500-1:1000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056730T
SDP
View Documents
Novus Biologicals
NB10056730SS
0.05 mL
Each for $206.50
Only null left
Add to Cart
 
NB10056730
SDP
View Documents
Novus Biologicals
NB10056730
0.2 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

TIRAP Monoclonal specifically detects TIRAP in Human samples. It is validated for Western Blot.
Specifications

Specifications

TIRAP
Western Blot 1:500-1:1000
Monoclonal
Ascites
RUO
P58753
114609
This antibody was developed against a synthetic peptide corresponding to a portion of human TIRAP.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
20D1055.1
Western Blot
Unconjugated
Mouse
Human
adapter protein wyatt, Adaptor protein Wyatt, MAL, MyD88 adapter-like protein, MyD88-2, TIR domain-containing adapter protein, toll/interleukin-1 receptor domain-containing adapter protein, toll-interleukin 1 receptor (TIR) domain containing adaptor protein, Toll-interleukin 1 receptor (TIR) domain-containing adaptor protein, Toll-like receptor adaptor protein, wyatt
TIRAP
IgM κ
Unpurified
In HUVEC, a 32 kDa band is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.