Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Antibody (619F7.06), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. DDX0470A488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0470A488 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0470A488 Supplier Novus Biologicals Supplier No. DDX0470A488100
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR3 Monoclonal specifically detects TLR3 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TLR3
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 619F7.06
Concentration 0.5mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Accession No. NP_003256
Gene Alias CD283, CD283 antigen, toll-like receptor 3
Gene Symbols TLR3
Host Species Mouse
Immunogen TLR3/CD283 was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7098
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.