Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAIL/TNFSF10 Antibody (RIK-2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10077865IR
Description
TRAIL/TNFSF10 Monoclonal specifically detects TRAIL/TNFSF10 in Human samples. It is validated for Flow Cytometry.Specifications
| TRAIL/TNFSF10 | |
| Monoclonal | |
| DyLight 755 | |
| Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| TRAIL (RIK-2) | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| RIK-2 | |
| Flow Cytometry | |
| TNFSF10 | |
| Human TRAIL-transfected mouse cell line | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Signal Transduction, Tumor Suppressors | |
| 8743 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction