Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF Antibody (VG1), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100664F
Description
VEGF Monoclonal specifically detects VEGF in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
VEGF | |
Monoclonal | |
FITC | |
P15692 | |
VEGFA | |
Recombinant VEGF 189 protein. | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
7422 | |
Human, Mouse, Rat, Canine | |
Purified |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
VG1 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This VEGF Antibody (VG1) detects the 189, 165 and 121 isoforms of VEGF | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction