Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mPER1 (residues 6-21) Rabbit anti-Mouse, Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB2201MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2201MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2201MI Supplier MilliporeSigma Supplier No. AB2201
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect mPER1 (residues 6-21) using this Anti-mPER1 (residues 6-21) Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mPER1 (residues 6-21)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum with 0.05% NaN3
Gene Symbols CAB2 ; KIAA0482 ; MGC88021; MGC9753; PER ; PERLD1; RIGUI ; hPER ; hPER1
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) BAA94086
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.