Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPO Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89024834 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-834 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-834 Supplier Abnova Corporation Supplier No. PAB14328

Rabbit polyclonal antibody raised against native MPO.

Myeloperoxidase (MPO) is a heme protein synthesized during myeloid differentiation that constitutes the major component of neutrophil azurophilic granules. Produced as a single chain precursor, myeloperoxidase is subsequently cleaved into a light and heavy chain. The mature myeloperoxidase is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme produces hypohalous acids central to the microbicidal activity of netrophils. [provided by RefSeq

Specifications

Antigen MPO
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native MPO.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from antiserum
Gene MPO
Gene Symbols MPO
Host Species Rabbit
Immunogen Native purified human polymorphonuclear leucocytes MPO.
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4353
Test Specificity This antiserum reacts with human myeloperoxidase.
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 250μL of deionized water, centrifuge to remove any insoluble material and store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.