Learn More
membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human MPP3 protein.
Supplier: Abnova Corporation H00004356D01P
Description
This gene product is a member of a family of membrane-associated proteins termed MAGUKs (membrane-associated guanylate kinase homologs). MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. This protein contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction. Alternatively spliced transcript variants have been identified. One transcript variant is experimentally supported, but it doesn't encode a protein. [provided by RefSeq]
Sequence: MPVLSEDSGLHETLALLTSQLRPDSNHKEEMGFLRDVFSEKSLSYLMKIHEKLRYYERQSPTPVLHSAVALAEDVMEELQAASVHSDERELLQLLSTPHLRAVLMVHDTVAQKNFDPVLPPLPDNIDEDFDEESVKIVRLVKNKEPLGATIRRDEHSGAVVVARIMRGGAADRSGLVHVGDELREVNGIAVLHKRPDEISQILAQSQGSITLKIIPATQEEDRLKESKVFMRALFHYNPREDRAIPCQEAGLPFQRRQVLEVVSQDDPTWWQAKRVGDTNLRAGLIPSKGFQERRLSYRRAAGTLPSPQSLRKPPYDQPCDKETCDCEGYLKGHYVAGLRRSFRLGCRERLGGSQEGKMSSGAESPELLTYEEVARYQHQPGERPRLVVLIGSLGARLHELKQKVVAENPQHFGVAVPHTTRPRKSHEKEGVEYHFVSKQAFEADLHHNKFLEHGEYKENLYGTSLEAIQAVMAKNKVCLVDVEPEALKQLRTSEFKPYIIFVKPAIQEKRKTPPMSPACEDTAAPFDEQQQEMAASAAFIDRHYGHLVDAVLVKEDLQGAYSQLKVVLEKLSKDTHWVPVSWVRSpecifications
membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human MPP3 protein. | |
MPP3 | |
DLG3 | |
Rabbit | |
Affinity Purified | |
RUO | |
4356 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001932.2 | |
MPP3 | |
MPP3 (NP_001923.2, 1 a.a. ∼ 585 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.