Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPP7, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89206984 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-206-984 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-206-984 Supplier Abnova Corporation Supplier No. H00143098K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human MPP7 peptide using ARM Technology.

Membrane-associated guanylate kinases (MAGUKs) are important adaptor proteins involved in the assembly of protein complexes at sites of cell-cell contact. They are found in synapses, adherens junctions, and tight junctions. All MAGUKs contain at least 1 PDZ domain, an SH3 domain, and a GUK domain, and many contain 1 or 2 L27 domains, which are involved in multimerization of MAGUKs. MPP7 belongs to the p55 stardust subfamily of MAGUKs, which is named for a Drosophila gene required for establishment of cell polarity in the developing fly embryo (Bohl et al., 2007 [PubMed 17237226]).[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen MPP7
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description membrane protein, palmitoylated 7 (MAGUK p55 subfamily member 7)
Formulation In 1X PBS, pH 7.4
Gene MPP7
Gene Accession No. MPP7
Gene Alias FLJ32798
Gene Symbols MPP7
Host Species Rabbit
Immunogen A synthetic peptide of human MPP7
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143098
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.