Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPPED2, Mouse, Clone: 2G1-1B7, Abnova™

Catalog No. 89001433 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-433 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-433 Supplier Abnova Corporation Supplier No. H00000744M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MPPED2.

This gene likely encodes a metallophosphoesterase. The encoded protein may play a role a brain development. Alternatively spliced transcript variants have been described. [provided by RefSeq

Sequence: MAHGIPSQGKVTITVDEYSSNPTQAFTHYNINQSRFQPPHVHMVDPIPYDTPKPAGHTRFVCISDTHSRTDGIQMPYGDILLHTGDFTELGLPSEVKKFNDWLGNLPYEYKIVIAGNHELTFDKEFMADLVKQDYYRFPSVSKLKPEDFDNVQSLLTNSIYLQDSEVTVKGFRIYGAPWTPWFNGWGFNLPRGQSLLDKWNLIPEGIDILMTHGPPLGFRDWVPKELQRVGCVELLNTVQRRVRPKLHVFGGIHEGYGIMTDGYTTYINASTCTVSFQPTNPPIIFDLPNPQGS

Specifications

Antigen MPPED2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G1-1B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant MPPED2.
Formulation ascites with no preservative
Gene MPPED2
Gene Accession No. BC031582
Gene Alias 239FB/C11orf8/D11S302E/FAM1B/Hs.46638/dJ1024C24.1/dJ873F21.1
Gene Symbols MPPED2
Host Species Mouse
Immunogen MPPED2 (AAH31582, 1 a.a. ∼ 294 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 744
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.