Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mps1 Mouse anti-Human, Clone: 3-472-1, MilliporeSigma™

Catalog No. 05682MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-682-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-682-MI Supplier MilliporeSigma Supplier No. 05682
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Mps1 with Anti-Mps1 Antibody, NT, clone 3-472-1 (Mouse Monoclonal Antibody), that has been shown to work in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mps1
Applications Western Blot
Classification Monoclonal
Clone 3-472-1
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl and 0.05% sodium azide.
Gene Accession No. NM_003318
Gene Symbols TTK, PYT, MPS1L1, ESK, FLJ38280
Host Species Mouse
Immunogen His-tagged fusion protein corresponding to amino acids 3-856 of human Mps1
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.