Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRAS, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002831 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-831 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-831 Supplier Abnova Corporation Supplier No. H00022808B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human MRAS protein.

Members of the RAS superfamily of GTP-binding proteins, which includes MRAS, are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. They are oncogenically activated in a significant fraction of tumors.[supplied by OMIM

Sequence: MATSAVPSDNLPTYKLVVVGDGGVGKSALTIQFFQKIFVPDYDPTIEDSYLKHTEIDNQWAILDVLDTAGQEEFSAMREQYMRTGDGFLIVYSVTDKASFEHVDRFHQLILRVKDRESFPMILVANKVDLMHLRKITREQGKEMATKHNIPYIETSAKDPPLNVDKAFHDLVRVIRQQIPEKSQKKKKKTKWRGDRATGTHKLQCVIL

Specifications

Antigen MRAS
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MRAS protein.
Formulation PBS with no preservative; pH 7.4
Gene MRAS
Gene Accession No. NM_012219
Gene Alias FLJ42964/M-RAs/R-RAS3/RRAS3
Gene Symbols MRAS
Host Species Mouse
Immunogen MRAS (NP_001078518.1, 1 a.a. ∼ 208 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22808
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.