Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mRNA-capping enzyme Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 061354
Description
Use Anti-mRNA-capping enzyme Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunocytochemistry to detect mRNA-capping enzyme also known as mRNA-capping enzyme, HCAP1, HCE.
Specifications
mRNA-capping enzyme | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
Immunocytochemistry, Western Blot | |
Unconjugated | |
RNGTT; CAP1A | |
GST-tagged recombinant protein corresponding to human mRNA-capping enzyme. | |
100 μL | |
NP_003791 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction