Learn More
MRP-1 Mouse anti-Human, Unlabeled, Clone: QCRL-3, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 557594
Description
Reacts with an intracellular epitope of the multidrug resistance protein (MRP1). MRP1 is a 190kDa integral membrane phosphoglycoprotein, member of the ATP-binding cassette transporter proteins, overexpressed in some drug-selected resistant cell lines and has been shown to cause multidrug resistance in transfected cells. Clone QCRL-3 was generated using non-denatured membranes from H69AR, an MRP1-overexpressing, multidrug resistant, drug-selected cell line. Its epitope has been localized to the first nucleotide binding domain of MRP1 between amino acids 617 and 932.3 QCRL-3 does not cross-react with human MDR1 or MDR3 gene products, nor with murine MRP1. It is reported that mAb QCRL-3 inhibits the ATP-dependent transport activity of MRP1 in inside-out membrane vesicles.
Host Species: Mouse
Clone: QCRL-3
Isotype: IgG2a, κ
Cross Reactivity: Human
Flow Cytometry

Specifications
MRP-1 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
QCRL-3 | |
Unconjugated | |
Mouse | |
0.1 mg | |
Cell Biology | |
Human | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.