Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRP-1 Mouse anti-Human, FITC, Clone: QCRL-3, BD

Catalog No. BDB557593 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557593 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557593 Supplier BD Biosciences Supplier No. 557593
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with an intracellular epitope of the multidrug resistance protein (MRP1). MRP1 is a 190kDa integral membrane phosphoglycoprotein, member of the ATP-binding cassette transporter proteins, overexpressed in some drug-selected resistant cell lines and has been shown to cause multidrug resistance in transfected cells. Clone QCRL-3 was generated using non-denatured membranes from H69AR, an MRP1-overexpressing, multidrug resistant, drug-selected cell line. Its epitope has been localized to the first nucleotide binding domain of MRP1 between amino acids 617 and 932.3 QCRL-3 does not cross-react with human MDR1 or MDR3 gene products, nor with murine MRP1. It is reported that mAb QCRL-3 inhibits the ATP-dependent transport activity of MRP1 in inside-out membrane vesicles.

Host Species: Mouse
Clone: QCRL-3
Isotype: IgG2a, κ
Cross Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP-1
Applications Flow Cytometry
Classification Monoclonal
Clone QCRL-3
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.