Learn More
anti-MRP1, Clone: IU2H10, Invitrogen™
Supplier: Thermo Scientific MA116823
Description
The MRP molecule, an integral membrane glycophosphoprotein belonging to the same superfamily of ATP-binding cassette transmembrane transporter proteins as P-glycoprotein, is overexpressed in many P-glycoprotein-negative, multidrug resistant cell lines and tumours. It is believed that the main function of MRP in drug resistance is that of a plasma membrane drug efflux pump.
Immunofluorescence, Western Blotting

Specifications
MRP1 | |
IU2H10 | |
Mouse | |
100 μL | |
Human, Mouse | |
IgG1 |
Monoclonal | |
Unconjugated | |
Recombinant peptide (a.a. 1-33) of human MRP1. | |
Primary | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.