Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRP1 Mouse anti-Human, Clone: MRPm5, MilliporeSigma™

Catalog No. MAB4147MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4147MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4147MI Supplier MilliporeSigma Supplier No. MAB4147
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect MRP1 using this Anti-MRP1 Antibody, clone MRPm5 validated for use in flow cytometry, western blot, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP1
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MRPm5
Conjugate Unconjugated
Formulation Tissue culture supernatant, serum-free. Liquid containing 0.7% BSA and 0.1% sodium azide.
Gene Accession No. NM_019901.1; NM_019902.1; NM_004996.3; NM_019862.2; NM_019898.2; NM_019899.2; NM_019900.2
Gene Symbols ABCC1, 4transporter), ABC29, GS-X, ABCC, MRP1, DKFZp781G125, MRP, DKFZp686N04233
Host Species Mouse
Immunogen Bacterial fusion protein of MRP1 containing amino acids 986-1204 of the protein.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.