Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-MRP1, Clone: QCRL-1, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB4100/DEL

Description
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting

Specifications
MRP1 | |
Unconjugated | |
Non-denatured membranes prepared from the human small cell lung cancer cell line, H69AR, which highly overexpresses MRP1 (Cole et al., 1992; Hipfner et al., 1994, 1999). | |
RUO | |
Reacts with a defined linear epitope (aa 918-924) of MRP1, a 190 kDa transmembrane, phosphoglycoprotein overexpressed in various human multidrug resistant tumor cell lines, and tumors (Cole et al., 1992; Hipfner et al., 1994, 1996, 1999; Wright et al, 199 | |
Human | |
IgG1 |
Monoclonal | |
Mouse | |
100 μg | |
Endocrine/Metabolism | |
Primary | |
Semi-purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction