Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-MRP1, Fluorescein, Clone: QCRL, R&D Systems™
Supplier: R&D Systems IC19291F
This item is not returnable.
View return policy
Description
Applications:
Flow Cytometry (Intracellular Staining)
Specifications
MRP1 | |
Monoclonal | |
Fluorescein | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
ABCC1 | |
H69AR human small cell lung cancer cell line membrane extracts | |
100 Tests | |
Primary | |
Detects human MRP1 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Flow Cytometry | |
QCRL | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
ABC29, ABCC, ABCC1, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4363 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction