Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRP2 Mouse anti-Human, Clone: M2 III-6, MilliporeSigma™

Catalog No. MAB4150MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4150MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4150MI Supplier MilliporeSigma Supplier No. MAB4150
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect MRP2 using this Anti-MRP2 Antibody, C-terminus, clone M2 III-6 validated for use in western blot, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP2
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone M2 III-6
Conjugate Unconjugated
Formulation Liquid containing 0.7% BSA and 0.1% sodium azide.
Gene Accession No. NM_000392.2
Gene Symbols ABCC2, MRP2, ABC#, CMOAT1, CMOAT, ABC30, CMRP, DJS, KIAA1010, cMRP
Host Species Mouse
Immunogen Bacterial fusion protein of MRP2 containing the 202-aa C-terminal end
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 6 months. −20°C for long term.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.